missing translation for 'onlineSavingsMsg'
Learn More

EHMT1/GLP1 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 09078MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09078MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09078MI Supplier MilliporeSigma Supplier No. 09078
Only null left

Rabbit Polyclonal Antibody

Use Anti-EHMT1/GLP1 Antibody (Rabbit Polyclonal Antibody) validated in western blotting to detect EHMT1/GLP1 also known as H3-K9-HMTase 5, Histone H3-K9 methyltransferase 5.

Specifications

Antigen EHMT1/GLP1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Immunoaffinity purified rabbit IgG in PBS containing 0.1% sodium azide, 30% glycerol
Gene Symbols EHMT1; EUHMTASE1; KMT1D; KIAA1876
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide (KHTQDSARVNPQDGTNTLTR(C) corresponding to a.a. 40-59 of human EMHT1/GLP1.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_012117.1; 79813
Target Species Human
Content And Storage -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles
Isotype IgG
Show More Show Less

For Research Use Only