Learn More
DnaJ (Hsp40) homolog, subfamily B, member 6 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human DNAJB6 protein.
Supplier: Abnova Corporation H00010049B01
Description
This gene encodes a member of the DNAJ protein family. DNAJ family members are characterized by a highly conserved amino acid stretch called the 'J-domain' and function as one of the two major classes of molecular chaperones involved in a wide range of cellular events, such as protein folding and oligomeric protein complex assembly. This family member may also play a role in polyglutamine aggregation in specific neurons. Alternative splicing of this gene results in multiple transcript variants; however, not all variants have been fully described. [provided by RefSeq]
Sequence: MVDYYEVLGVQRHASPEDIKKAYRKLALKWHPDKNPENKEEAERKFKQVAEAYEVLSDAKKRDIYDKYGKEGLNGGGGGGSHFDSPFEFGFTFRNPDDVFREFFGGRDPFSFDFFEDPFEDFFGNRRGPRGSRSRGTGSFFSAFSGFPSFGSGFSSFDTGFTSFGSLGHGGLTSFSSTSFGGSGMGNFKSISTSTKMVNGRKITTKRIVENGQERVEVEEDGQLKSLTINGVADDDALAEERMRRGQNALPAQPAGLRPPKPPRPASLLRHAPHCLSEEEGEQDRPRAPGPWDPLASAAGLKEGGKRKKQKQREESKKKKSTKGNHSpecifications
| DnaJ (Hsp40) homolog, subfamily B, member 6 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human DNAJB6 protein. | |
| DNAJB6 | |
| DJ4/DKFZp566D0824/DnaJ/FLJ42837/HHDJ1/HSJ-2/HSJ2/MGC1152/MGC117297/MRJ/MSJ-1 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 10049 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| No additive | |
| NM_058246.3 | |
| DNAJB6 | |
| DNAJB6 (NP_490647.1, 1 a.a. ∼ 326 a.a) full-length human protein. | |
| RUO | |
| Primary | |
| Human | |
| Serum |