missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA/Histone H1 Mouse anti-All, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB3864
Description
Anti-DNA/Histone H1 Antibody is a high quality Mouse Monoclonal Antibody for the detection of DNA/Histone H1 and has been validated in ELISA, flow cytometry, immunocytochemistry and immunohistochemistry.Specifications
| DNA/Histone H1 | |
| Monoclonal | |
| Protein A purified culture supernatant. Liquid in PBS | |
| HIST1H1E, H1.4, dJ221C16.5, H1F4, MGC116819 | |
| Human Raji cell nuclei | |
| Primary | |
| −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. | |
| IgG2a |
| ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry | |
| Unconjugated | |
| NM_005321.2 | |
| Mouse | |
| 100 μL | |
| All species | |
| Purified |
For Research Use Only