missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl-Histone H4 (Lys20) Mouse anti-Bovine, Human, Mouse, Rat, Clone: 6G7/H4, MilliporeSigma™

Catalog No. 05672MI
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05672MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05672MI Supplier MilliporeSigma Supplier No. 05672
Only null left

Mouse Monoclonal Antibody

Use Anti-dimethyl-Histone H4 (Lys20) Antibody, clone 6G7/H4 (mouse monoclonal antibody) validated in immunoprecipitation, western blot, immunohistochemistry to detect dimethyl-Histone H4 (Lys20) also known as H4K20me2, Histone H4 (di methyl K20).

Specifications

Antigen dimethyl-Histone H4 (Lys20)
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 6G7/H4
Conjugate Unconjugated
Formulation PBS
Gene Accession No. NM_175054
Gene Symbols HIST1H4A, H4/J, HIST1H4F, HIST1H4L, H4FN, H4FA, H4FH, HIST1H4H, H4F2, H4FM, H4/A, H4FK, H4FG, HIST2H4, H4FB, H4/K, HIST1H4I, H4/N, HIST1H4B, H4FE, H4/M, H4/a, HIST1H4E, HIST4H4, HIST2H4A, H4FC, H4FI, H4/H, HIST1H4C, H4/G, HIST1H4K, H4FJ, H4FD, H4/I, H4/B;
Host Species Mouse
Immunogen peptide corresponding to amino acids 17-28 of human histone H4 (RHRdimKVLRDNIQGC) dimethylated on lysine 20.
Purification Method Protein G chromatography
Quantity 200 μg
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Rat
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only