Learn More
Dimethyl-Histone H3 (Lys27) Rabbit anti-Vertebrates, Clone: 614M, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier:  MilliporeSigma 04821
Catalog No. 04821MI
Only null left
Description
Description
Anti-dimethyl-Histone H3 (Lys27) Antibody, clone 614M is a rabbit monoclonal antibody for detection of dimethyl-Histone H3 (Lys27) also known as H3K27me2, Histone H3 (di methyl K27) and has been validated in western blot, PIA.
Specifications
Specifications
dimethyl-Histone H3 (Lys27)
Monoclonal
Unconjugated
NM_021059.2
Peptide containing the sequence (ARme2KSA) in which me2 corresponds to dimethyl lysine at residue 27 of human histone H3.
100 μL
H3, H3.2, H3/M, H3/m, H3/o, H3F2, H3FM, MGC9629, OTTHUMP00000014041
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
IgG
Inhibition Assays, Western Blot
614M
Cultured supernantant in 0.05% sodium azide.
Rabbit
Unpurified supernatant
Primary
Vertebrates
Supernatant
Product Suggestions
Product Suggestions