missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-dimethyl-Histone H3 (Arg17), Polyclonal, MilliporeSigma™
Supplier: MilliporeSigma 07214
Specifications
| dimethyl-Histone H3 (Arg17) | |
| Polyclonal | |
| Purified rabbit polyclonal IgG in buffer containing 0.14M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide, 3.5mg/mL BSA and 30% glycerol. | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Human, Vertebrates | |
| IgG |
| ELISA, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| HIST3H3; H3FT; MGC126886; H3t; MGC126888; H3T; H3/g; H3.4; H3/t | |
| BSA-conjugated, synthetic peptide (TGGKAPMe2RKQLA-C) corresponding to amino acids 11-21 of human Histone H3. | |
| 100 μL | |
| NP_003484 | |
| −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. |
For Research Use Only