Learn More
Dimethyl Histone H2B Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABE353
Description
This Anti-dimethyl Histone H2B Antibody (Pro1) is validated for use in Western Blotting and Immunocytochemistry and Dot Blot for the detection of dimethyl Histone H2B (Pro1).Specifications
| dimethyl Histone H2B (Pro1) | |
| Polyclonal | |
| Purified rabbit Polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Human, Mouse |
| Dot Blot, Immunocytochemistry, Western Blot | |
| Unconjugated | |
| HIST1H2BC; H2BFL; HIST1H2BE; H2BFH; HIST1H2BF; H2BFG; HIST1H2BG; H2BFA; HIST1H2BI; H2BFK | |
| KLH-conjugated linear peptide corresponding to the N-terminus of Human dimethyl Histone H2B methylated at Pro1. | |
| 100 μg | |
| NP_003509 | |
| 2-8°C for 12 months |
For Research Use Only