missing translation for 'onlineSavingsMsg'
Learn More

DFF40 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB16926MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB16926MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB16926MI Supplier MilliporeSigma Supplier No. AB16926
Only null left

Rabbit Polyclonal Antibody

Detect DFF40 using this Anti-DFF40 Antibody validated for use in western blotting.

Specifications

Antigen DFF40
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified immunoglobulin. Liquid in PBS containing 0.02% sodium azide.
Gene Symbols DFFB; CAD; DFF40; DFF-40; CPAN; DFF2; EC 3.-.-.-
Host Species Rabbit
Immunogen A peptide corresponding to amino acids 147-164 of mouse CAD (Enari et al. 1998). The sequence differs from human DFF40 by two amino acids (Halenbeck et al. 1998; Liu et al. 1998).
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001004285.1; NM_001004286.1; NM_004402.2
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 12 months
Form Purified
Show More Show Less

For Research Use Only