missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Cytokeratin 7 Mouse anti-Human, FITC, Clone: LP5K
Description
Detect Cytokeratin 7 using this Anti-Cytokeratin 7 Antibody, clone LP5K, FITC conjugated validated for use in immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin), flow cytometry.
Specifications
Specifications
| Antigen | Cytokeratin 7 |
| Applications | Flow Cytometry, Immunocytochemistry, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | LP5K |
| Conjugate | FITC |
| Formulation | The antibody is ammonium sulphate precipitation followed by ion exchange chromatography. The conjugate is supplied in phosphate buffered saline containing 10mM sodium azide and 1mg/ml bovine serum albumin. |
| Gene Accession No. | NM_005556.3 |
| Gene Symbols | KRT7, Sarcolectin, Keratin-7, K7, MGC3625, CK-7, MGC129731, SCL, Cytokeratin-7, K2C7, CK7, sarcolectin |
| Host Species | Mouse |
| Immunogen | Cytoskeletal proteins from cultured HeLa cells |
| Show More |
For Research Use Only