missing translation for 'onlineSavingsMsg'
Learn More

Cyclin A Mouse anti-Bovine, Human, Mouse, Mink, Clone: E67.1, MilliporeSigma™

Catalog No. MAB3682MI
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3682MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3682MI Supplier MilliporeSigma Supplier No. MAB3682
Only null left

Mouse Monoclonal Antibody

Anti-Cyclin A Antibody, clone E67.1 is a high quality Mouse Monoclonal Antibody for the detection of Cyclin A and has been validated in immunoprecipitation and western blotting.

Specifications

Antigen Cyclin A
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone E67.1
Conjugate Unconjugated
Formulation Ascites fluid. Liquid. Contains no preservative.
Gene Accession No. NM_001111045.1; NM_001111046.1; NM_001111047.1; NM_003914.3
Gene Symbols CCNA1, Cyclin-A1.
Host Species Mouse
Immunogen Bovine Cyclin A.
Quantity 100 μL
Primary or Secondary Primary
Target Species Bovine, Human, Mink, Mouse
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG2a
Show More Show Less

For Research Use Only