missing translation for 'onlineSavingsMsg'
Learn More

Cyclin A Mouse anti-Bovine, Human, Mouse, Clone: E23.1, MilliporeSigma™

Catalog No. MAB3680MI
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3680MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3680MI Supplier MilliporeSigma Supplier No. MAB3680
Only null left

Mouse Monoclonal Antibody

Detect Cyclin A with Anti-Cyclin A Antibody, clone E23.1 (Mouse Monoclonal Antibody), that has been shown to work in western blot, immunocytochemistry.

Specifications

Antigen Cyclin A
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone E23.1
Conjugate Unconjugated
Formulation Ascites fluid. Liquid. Contains no preservative.
Gene Accession No. NM_001111045.1; NM_001111046.1; NM_001111047.1; NM_003914.3
Gene Symbols CCNA1, Cyclin-A1.
Host Species Mouse
Immunogen Bovine Cyclin A fusion protein.
Quantity 100 μL
Primary or Secondary Primary
Target Species Bovine, Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG2a
Show More Show Less

For Research Use Only