Learn More
CPT1A, Mouse, Clone: 1D3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CPT1A.
Supplier: Abnova Corporation H00001374M02
Description
The mitochondrial oxidation of long-chain fatty acids is initiated by the sequential action of carnitine palmitoyltransferase I (which is located in the outer membrane and is detergent-labile) and carnitine palmitoyltransferase II (which is located in the inner membrane and is detergent-stable), together with a carnitine-acylcarnitine translocase. CPT I is the key enzyme in the carnitine-dependent transport across the mitochondrial inner membrane and its deficiency results in a decreased rate of fatty acid beta-oxidation. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: VFKNGKMGLNAEHSWADAPIVAHLWEYVMSIDSLQLGYAEDGHCKGDINPNIPYPTRLQWDIPGECQEVIETSLNTANLLANDVDFHSFPSpecifications
| CPT1A | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001876 | |
| CPT1A | |
| CPT1A (NP_001867.2, 461 a.a. ∼ 550 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 1D3 | |
| Mouse monoclonal antibody raised against a partial recombinant CPT1A. | |
| CPT1A | |
| CPT1/CPT1-L/L-CPT1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1374 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |