missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Chondroitin Sulfate D Monoclonal Antibody (MO-225) , TCI America™
Anti-Chondroitin Sulfate D Monoclonal Antibody (MO-225), TCI America™
Supplier: TCI America A28721VIAL
Specifications
| Chondroitin Sulfate D | |
| Monoclonal | |
| Mouse | |
| 1 Vial | |
| Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing. | |
| IgM κ |
| Immunohistochemistry, ELISA, Western Blot | |
| MO-225 | |
| Chick embryo limb bud proteoglycan (PG-M/versican) | |
| Primary | |
| Purified |