missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Anti-Chondroitin Sulfate A Monoclonal Antibody (LY111) , TCI America™
Specifications
Specifications
| Antigen | Chondroitin Sulfate A |
| Applications | Immunohistochemistry, ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | LY111 |
| Host Species | Mouse |
| Immunogen | Chick embryo limb bud proteoglycan (PG-M/versican) |
| Quantity | 1 Vial |
| Primary or Secondary | Primary |
| Content And Storage | Store at -20°C to -80°C (preferably at -80°C). It is not recommended storing at 4°C. Avoid repeated freezing and thawing. |
| Form | Purified |
| Show More |