missing translation for 'onlineSavingsMsg'
Learn More

chondroadherin, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant CHAD.

Supplier:  Abnova Corporation H00001101A01

Catalog No. 89-004-063


Only null left
Explore available promotions

Description

Description

Chondroadherin is a cartilage matrix protein thought to mediate adhesion of isolated chondrocytes. The protein contains 11 leucine-rich repeats flanked by cysteine-rich regions. The chondroadherin messenger RNA is present in chondrocytes at all ages. [provided by RefSeq

Sequence: LDNTNLEKFSDGAFLGVTTLKHVHLENNRLNQLPSNFPFDSLETLALTNNPWKCTCQLRGLRRWLEAKASRPDATCASPAKFKGQHIRDTDAFRSCKFPTKRSKKAGRH
Specifications

Specifications

chondroadherin
Polyclonal
Mouse polyclonal antibody raised against a partial recombinant CHAD.
CHAD
SLRR4A
Mouse
50 μL
Yes
1101
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
Unconjugated
50% glycerol
NM_001267
CHAD
CHAD (NP_001258, 251 a.a. ∼ 359 a.a) partial recombinant protein with GST tag.
RUO
Primary
Human
Serum
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions