missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDCP1 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABT180
Description
Anti-CDCP1 Antibody is a highly specific rabbit polyclonal antibody, that targets CUB domain-containing protein 1 and has been tested in western blotting and immunohistochemistry (Paraffin).Specifications
| CDCP1 | |
| Polyclonal | |
| Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Human |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| CDCP1; TRASK | |
| KLH-conjugated linear peptide corresponding to the cytoplasmic domain of human CDCP1. | |
| 100 μg | |
| NP_073753 | |
| 2-8°C for 12 months |
For Research Use Only