missing translation for 'onlineSavingsMsg'
Learn More

Cdc20 Mouse anti-Human, Clone: AR12, MilliporeSigma™

Catalog No. MAB3775MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3775MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3775MI Supplier MilliporeSigma Supplier No. MAB3775
Only null left

Mouse Monoclonal Antibody

Anti-Cdc20 Antibody, clone AR12 is a Mouse Monoclonal Antibody for detection of Cdc20 also known as Cell Division Cycle Protein 20 Homolog p55cdc and has been validated in IF and immunoprecipitation.

Specifications

Antigen Cdc20
Applications Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone AR12
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.02M phosphate buffer, 0.25M NaCl, pH 7.6, with 0.1% sodium azide.
Gene Accession No. NM_001255.2
Gene Symbols CDC20, CDC20A, MGC102824, p55CDC, P55CDC-LSB, bA276H19.3
Host Species Mouse
Immunogen Human Cdc20 recombinant protein
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only