missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ anti-CD8a PE, Clone: 53-6.7,
Rat Monoclonal Antibody
$224.35
Specifications
| Antigen | CD8a |
|---|---|
| Clone | 53-6.7 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
MABF1543MI
|
MilliporeSigma™
MABF1543 |
25 μg |
Each for $224.35
|
|
|||||
Description
Specifically detects CD8a Clone: 53-6.7 in Mouse samples, and it is validated for Flow Cytometry
The 53-6.7 antibody reacts with the 32-34kDa alpha subunit of mouse CD8, known as CD8a or CD8 alpha. CD8a can form a homodimer (CD8 alpha-alpha), but is more commonly expressed as a heterodimer with a second chain known as CD8b or CD8 beta. CD8 acts as a co-receptor in antigen recognition and subsequent T cell activation that is initiated upon binding of the T cell receptor (TCR) to antigen-bearing MHC Class I molecules. The cytoplasmic domains of CD8 provide binding sites for the tyrosine kinase lck, facilitating intracellular signaling events that lead to T cell activation, development, and cytotoxic effector functions. CD8+ cytotoxic T cells (CTLs) play an important role in inducing cell death of tumor cells, as well as cells infected by virus, bacteria or parasites.The 53-6.7 antibody is widely used as a phenotypic marker for mouse CD8a expression on cytotoxic T cells, thymocytes, as well as on certain cell types that do not also express the TCR, including some NK cells and lymphoid dendritic cells.Specifications
| CD8a | |
| Flow Cytometry | |
| PE | |
| Rat | |
| Inflammation, Immunology | |
| Purified Rat monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2. | |
| CD8a | |
| IgG2a κ | |
| Purified by size-exclusion chromatography |
| 53-6.7 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse | |
| P01731 | |
| Proprietary immunogen | |
| Primary | |
| 2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze |
For Research Use Only