missing translation for 'onlineSavingsMsg'
Learn More

CD68 molecule, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a partial recombinant CD68.

Supplier:  Abnova Corporation H00000968A01

Catalog No. 89004055


Only null left

Description

Description

This gene encodes a 110kDa transmembrane glycoprotein that is highly expressed by human monocytes and tissue macrophages. It is a member of the lysosomal/endosomal-associated membrane glycoprotein (LAMP) family. The protein primarily localizes to lysosomes and endosomes with a smaller fraction circulating to the cell surface. It is a type I integral membrane protein with a heavily glycosylated extracellular domain and binds to tissue- and organ-specific lectins or selectins. The protein is also a member of the scavenger receptor family. Scavenger receptors typically function to clear cellular debris, promote phagocytosis, and mediate the recruitment and activation of macrophages. Alternative splicing results in multiple transcripts encoding different isoforms. [provided by RefSeq

Sequence: NGSQPCVHLQAQIQIRVMYTTQGGGEAWGISVLNPNKTKVQGSCEGAHPHLLLSFPYGHLSFGFMQDLQQKVVYLSYMAVEYNVSFPHAAKWTFSAQNA
Specifications

Specifications

CD68 molecule
Polyclonal
Mouse polyclonal antibody raised against a partial recombinant CD68.
CD68
DKFZp686M18236/GP110/SCARD1
Mouse
50 μL
Yes
968
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
ELISA, Western Blot
Unconjugated
50% glycerol
NM_001251
CD68
CD68 (NP_001242, 164 a.a. ∼ 262 a.a) partial recombinant protein with GST tag.
RUO
Primary
Human
Serum
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions