missing translation for 'onlineSavingsMsg'
Learn More

CD6 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABN168MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN168MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN168MI Supplier MilliporeSigma Supplier No. ABN168
Only null left

Rabbit Polyclonal Antibody

Anti-CD6 Antibody is a highly specific rabbit polyclonal antibody, that targets CD6 and has been tested in immunohistochemistry.

Specifications

Antigen CD6
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols CD6
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human CD6.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_006716
Target Species Human
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only