missing translation for 'onlineSavingsMsg'
Learn More

CD25 Mouse anti-Human, FITC, Clone: ACT-1, MilliporeSigma™

Catalog No. FCMAB189FMI
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB189FMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB189FMI Supplier MilliporeSigma Supplier No. FCMAB189F
Only null left

Mouse Monoclonal Antibody

This Milli-Mark™ Anti-CD25-FITC Antibody, clone ACT-1 is validated for use in FC for the detection of CD25.

Specifications

Antigen CD25
Applications Flow Cytometry
Classification Monoclonal
Clone ACT-1
Conjugate FITC
Formulation Purified Mouse monoclonal IgG1 conjugated to FITC in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.
Gene Accession No. NP_000408
Host Species Mouse
Immunogen Phytohemagglutinin-stimulated blood lymphocytes
Purification Method Protein A Purified
Quantity 100 Tests
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from light.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only