missing translation for 'onlineSavingsMsg'
Learn More

CD152 (CTLA-4) Armenian Hamster anti-Mouse, APC, Clone: UC10-4F10-11, MilliporeSigma™

Catalog No. mabf389mi
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF389MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF389MI Supplier MilliporeSigma Supplier No. MABF389
Only null left

Armenian Hamster Monoclonal Antibody

This armenian hamster monoclonal antibody Anti-CD152 (CTLA-4) (mouse), APC and clone UC10-4F10-11 validated for use in Flow Cytometry for the detection of CD152 (CTLA-4).

Specifications

Antigen CD152 (CTLA-4)
Applications Flow Cytometry
Classification Monoclonal
Clone UC10-4F10-11
Concentration 0.2mg/mL
Conjugate APC
Formulation Purified Armenian Hamster Monoclonal Antibody in buffer containing 10mM NaH2PO4, 150mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide.
Gene Symbols Ctla4, Cd152
Host Species Armenian Hamster
Immunogen Proprietary immunogen
Purification Method Size Exclusion Chromatography
Quantity 100 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C, protect from prolonged exposure to light. Do not Freeze.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only