missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ anti-CD11c Red Fluor 710, Clone: N418,

Armenian Hamster Monoclonal Antibody

$262.46 - $444.46

Specifications

Antigen CD11c
Clone N418
Applications Flow Cytometry
Classification Monoclonal
Conjugate Red Fluor 710
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
MABF1501MI
View Documents
MilliporeSigma™
MABF1501
25 μg
Each for $262.46
Only null left
 
MABF1502MI
View Documents
MilliporeSigma™
MABF1502
100 μg
Each for $444.46
Only null left
 
Description

Description

Specifically detects CD11c Clone: N418 in Mouse samples, and it is validated for Flow Cytometry

The N418 antibody reacts with mouse CD11c, also known as integrin alpha X. This 150kDa cell surface glycoprotein is part of a family of integrin receptors that mediate adhesion between cells (cell-cell) and components of the extracellular matrix, e.g. fibrinogen (cell-matrix). In addition, integrins are active signaling receptors which recruit leukocytes to inflammatory sites and promote cell activation. Complete, functional integrin receptors consist of distinct combinations of integrin chains which are differentially expressed. Integrin alpha X (CD11c) assembles with Integrin beta-2 (CD18) into a receptor complex known as CR4 which can bind and induce signaling through ICAMs and VCAM-1 on endothelial cells and can also facilitate removal of iC3b bearing foreign cells.The N418 antibody is widely used as a marker for CD11c expression on dendritic cells (DC), often in parallel with markers for CD11b, for identification of developmental stages and mature subsets of this cell type. CD11c is prominently expressed on tissue macrophages, and is also detected on some types of activated T cells and intestinal intraepithelial lymphocytes (IEL).
Specifications

Specifications

CD11c
Flow Cytometry
Red Fluor 710
Armenian Hamster
Inflammation, Immunology
Purified Armenian Hamster monoclonal containing 10mM NaH2PO4, 150mM NaCl, 0.09% NaN3, 0.1% gelatin, pH 7.2.
Itgax
IgG
Purified by size-exclusion chromatography
N418
Monoclonal
Purified
RUO
Mouse
Q9QXH4
Proprietary immunogen
Primary
2°C to 8°C undiluted for one year from date of shipment protected from light; Do not freeze
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only