missing translation for 'onlineSavingsMsg'
Learn More

CD11b Rat anti-Human, Mouse, PE-Cy5, Clone: M1/70, MilliporeSigma™

Catalog No. MABF515MI
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF515MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF515MI Supplier MilliporeSigma Supplier No. MABF515
Only null left

Rat Monoclonal Antibody

This rat monoclonal antibody Anti-CD11b (human/mouse), PE-Cy5, clone M1/70 validated for use in immunoprecipitation, immunohistochemistry, Flow Cytometry and Immunofluorescence for the detection of CD11b.

Specifications

Antigen CD11b
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone M1/70
Concentration 0.2mg/mL
Conjugate PE-Cyanine5
Formulation Purified Rat Monoclonal Antibody in buffer containing 10mM NaH2PO4, 150mM NaCl, 0.1% gelatin, pH 7.2, and up to 0.1% sodium azide.
Gene Symbols ITGAM, CD11B, CR3A
Host Species Rat
Immunogen Proprietary immunogen
Purification Method Size Exclusion Chromatography
Quantity 25 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C, protect from prolonged exposure to light. Do not Freeze.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only