missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCT-beta Mouse anti-Human, Clone: F39 P7 F11, MilliporeSigma™
Description
This Anti-CCT-beta Antibody, clone F39 P7 F11 is validated for use in western blotting for the detection of CCT-beta.
Specifications
Specifications
| Antigen | CCT-beta |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | F39 P7 F11 |
| Conjugate | Unconjugated |
| Formulation | Purified mouse monoclonal IgG2aκ in buffer containing PBS and 0.05% sodium azide. |
| Gene Accession No. | NP_006422 |
| Gene Symbols | PCYT1B, CCTB |
| Host Species | Mouse |
| Immunogen | Synthetic Peptide corresponding to the C-Terminus of Human CCT-beta |
| Show More |
For Research Use Only