missing translation for 'onlineSavingsMsg'
Learn More

carbonyl reductase 3 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89004051
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89004051 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89004051 Supplier Abnova Corporation Supplier No. H00000874A01
Only null left

Mouse polyclonal antibody raised against a partial recombinant CBR3.

Carbonyl reductase 3 catalyzes the reduction of a large number of biologically and pharmacologically active carbonyl compounds to their corresponding alcohols. The enzyme is classified as a monomeric NADPH-dependent oxidoreductase. CBR3 contains three exons spanning 11.2 kilobases and is closely linked to another carbonyl reductase gene - CBR1. [provided by RefSeq

Sequence: KNEVHEREGWPNSPYGVSKLGVTVLSRILARRLDEKRKADRILVNACCPGPVKTDMDGKDSIRTVEEGAETPVYLALLPPDATEPQGQLVHDKVVQNW

Specifications

Antigen carbonyl reductase 3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant CBR3.
Formulation 50% glycerol
Gene CBR3
Gene Accession No. NM_001236
Gene Alias SDR21C2/hCBR3
Gene Symbols CBR3
Host Species Mouse
Immunogen CBR3 (NP_001227, 180 a.a. ∼ 277 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 874
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less