missing translation for 'onlineSavingsMsg'
Learn More
Learn More
carbonyl reductase 1, Mouse, Clone: 4D12-1G8, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant CBR1.
Supplier: Abnova Corporation H00000873M02
Description
Carbonyl reductase is one of several monomeric, NADPH-dependent oxidoreductases having wide specificity for carbonyl compounds. This enzyme is widely distributed in human tissues. Another carbonyl reductase gene, CRB3, lies close to this gene on chromosome 21q. [provided by RefSeq
Sequence: MSSGIHVALVTGGNKGIGLAIVRDLCRLFSGDVVLTARDVTRGQAAVQQLQAEGLSPRFHQLDIDDLQSIRALRDFLRKEYGGLDVLVNNAGIAFKVADPTPFHIQAEVTMKTNFFGTRDVCTELLPLIKPQGRVVNVSSIMSVRALKSCSPELQQKFRSETITEEELVGLMNKFVEDTKKGVHQKEGWPSSAYGVTKIGVTVLSRIHARKLSEQRKGDKILLNACCPGWVRTDMAGPKATKSPEEGAETPVYLALLPPDAEGPHGQFVSEKRVEQWSpecifications
| carbonyl reductase 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC002511 | |
| CBR1 | |
| CBR1 (AAH02511.1, 1 a.a. ∼ 277 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Stem Cell Biology | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 4D12-1G8 | |
| Mouse monoclonal antibody raised against a full length recombinant CBR1. | |
| CBR1 | |
| CBR/SDR21C1/hCBR1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 873 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |