missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase3 (active form) Mouse anti-Human, Clone: 3D9.3, MilliporeSigma™
Description
Anti-Caspase3 (active form), clone 3D9.3 is an antibody targeting the Caspase3 (active form) protein, validated for use in western blot, immunocytochemistry and Flow.
Specifications
Specifications
| Antigen | Caspase3 (active form) |
| Applications | Flow Cytometry, Immunocytochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 3D9.3 |
| Conjugate | Unconjugated |
| Formulation | Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide. |
| Gene Accession No. | NP_004337 |
| Gene Symbols | CASP3, CPP32 |
| Host Species | Mouse |
| Immunogen | KLH-conjugated linear peptide corresponding to human Caspase3 (active form). |
| Show More |
For Research Use Only