missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caspase 9 Mouse anti-Human, Clone: 96-2-22, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05572
Description
This Anti-Caspase 9 Antibody, clone 96-2-22 is validated for use in western blotting for the detection of Caspase 9.Specifications
| Caspase 9 | |
| Monoclonal | |
| Unconjugated | |
| NM_032996 | |
| Mouse | |
| Protein G purfied | |
| Primary | |
| −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| 96-2-22 | |
| Protein A Purified immunoglobulin in Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105MNaCl, 0.035% sodium azide as a preservative. | |
| CASP9, CASP-9, ICE-LAP6, MCH6, APAF3, CASPASE-9c, OTTHUMP00000044594, APAF-3 | |
| The N-terminal fragment of human Caspase 9 (amino acid residues 1-134) | |
| 100 μg | |
| Human | |
| Purified |
For Research Use Only