missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CART, Mouse, Clone: 3E4, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant CART.
Supplier: Abnova Corporation H00009607M01
Description
CART appears to have roles in reward, feeding, and stress, and it has the functional properties of an endogenous psychostimulant (Kuhar et al., 2002 [PubMed 12147208]).[supplied by OMIM
Sequence: MESSRVRLLPLLGAALLLMLPLLGTRAQEDAELQPRALDIYSAVDDASHEKELIEALQEVLKKLKSKRVPIYEKKYGQVPMCDAGEQCAVRKGARIGKLCDCPRGTSCNSFLLKCLSpecifications
| CART, | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC029882 | |
| CARTPT | |
| CART (AAH29882, 1 a.a. ∼ 116 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 3E4 | |
| Mouse monoclonal antibody raised against a full-length recombinant CART. | |
| CARTPT | |
| CART | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9607 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |