missing translation for 'onlineSavingsMsg'
Learn More

CAPG (M01A), Mouse anti-Human, Clone: 1F1, Abnova™

Mouse monoclonal antibody raised against a full-length recombinant CAPG.

Supplier:  Abnova Corporation H00000822M01A

Catalog No. 89-016-424


Only null left
Explore available promotions

Description

Description

This gene encodes a member of the gelsolin/villin family of actin-regulatory proteins. The encoded protein reversibly blocks the barbed ends of F-actin filaments in a Ca2+ and phosphoinositide-regulated manner, but does not sever preformed actin filaments. By capping the barbed ends of actin filaments, the encoded protein contributes to the control of actin-based motility in nonmuscle cells. Alternatively spliced transcript variants have been observed, but have not been fully described. [provided by RefSeq]

Sequence: MYTAIPQSGSPFPGSVQDPGLHVWRVEKLKPVPVAQENQGVFFSGDSYLVLHNGPEEVSHLHLWIGQQSSRDEQGACAVLAVHLNTLLGERPVQHREVQGNESDLFMSYFPRGLKYQEGGVESAFHKTSTGAPAAIKKLYQVKGKKNIRATERALNWDSFNTGDCFILDLGQNIFAWCGGKSNILERNKARDLALAIRDSERQGKAQVEIVTDGEEPAEMIQVLGPKPALKEGNPEEDLTADKANAQAAALYKVSDATGQMNLTKVADSSPFALELLISDDCFVLDNGLCGKIYIWKGRKANEKERQAALQVAEGFISRMQYAPNTQVEILPQGRESPIFKQFFKDWK
Specifications

Specifications

CAPG
Monoclonal
Unconjugated
ascites with no preservative
BC000728
CAPG
CAPG (AAH00728.1, 1 a.a. ∼ 348 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
RUO
822
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Liquid
ELISA, Western Blot
1F1
Mouse monoclonal antibody raised against a full-length recombinant CAPG.
CAPG
AFCP/MCP
Mouse
200 μL
Primary
Human
Antibody
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions