Learn More
CALML5, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CALML5.
Supplier: Abnova Corporation H00051806A01
Description
This gene encodes a novel calcium binding protein expressed in the epidermis and related to the calmodulin family of calcium binding proteins. Functional studies with recombinant protein demonstrate it does bind calcium and undergoes a conformational change when it does so. Abundant expression is detected only in reconstructed epidermis and is restricted to differentiating keratinocytes. In addition, it can associate with transglutaminase 3, shown to be a key enzyme in the terminal differentiation of keratinocytes. [provided by RefSeq
Sequence: AQLRKLISEVDSDGDGEISFQEFLTAARKARAGLEDLQVAFRAFDQDGDGHITVDELRRAMAGLGQPLPQEELDAMIREADVDQDGRVNYEEFARMLAQESpecifications
| CALML5 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant CALML5. | |
| CALML5 | |
| CLSP | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_017422 | |
| CALML5 | |
| CALML5 (NP_059118, 47 a.a. ∼ 146 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 51806 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |