missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcineurin Aα Rabbit anti-Mammals, Human, Rat, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 071492
Description
This Anti-Calcineurin Aα Antibody is validated for use in western blotting for the detection of Calcineurin Aα.Specifications
| Calcineurin Aα | |
| Polyclonal | |
| Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH7.4) 150mM NaCl with 40% gylerol. Contains no preservative. | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Human, Mammalia, Mouse, Rat |
| Western Blot | |
| Unconjugated | |
| MGC138448; NF-ATC ; NF-ATc ; NF-ATc1 ; NFAT2 ; NFATC; NFATc | |
| KLH-conjugagted synthetic peptide corresponding to Calcineurin Aα. | |
| 100 μL | |
| NP_000935.1 | |
| -20°C in undiluted aliquots for up to 12 months, Avoid Freeze/Thaw Cycles |
For Research Use Only