missing translation for 'onlineSavingsMsg'
Learn More
Learn More
chromosome 15 open reading frame 29, Mouse, Clone: 1E7, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant C15orf29.
Supplier: Abnova Corporation H00079768M01A
Description
Sequence: MASETHNVKKRNFCNKIEDHFIDLPRKKISNFTNKNMKEVKKSPKQLAAYINRTVGQTVKSPDKLRKVIYRRKKVHHPFPNPCYRKKQSPGSGGCDMANKENELACAGHLPEKLHHDSRTYLVNSSDSGSSQTESPSSKYSGFFSEVSQDHETMAQVLFSRNMRLNVALTFWRKRSISELVAYLLRIEDLGVVVDCLPVLTNCLQEEKQYISLGCCVDLLPLVKSLLKSKFEEYVIVGLNWLQAVIKRWWSELSSKTEIINDGNIQILKQQLSGLWEQENHLTLVPGYTGNIAKDVDAYLLQLHSpecifications
| chromosome 15 open reading frame 29 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC026234 | |
| C15orf29 | |
| C15orf29 (AAH26234.1, 1 a.a. ∼ 304 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
| ELISA, Western Blot | |
| 1E7 | |
| Mouse monoclonal antibody raised against a full length recombinant C15orf29. | |
| C15orf29 | |
| FLJ22557/MGC117356 | |
| Mouse | |
| 200 μL | |
| Yes | |
| 79768 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |