missing translation for 'onlineSavingsMsg'
Learn More

Brain-Specific Angiogenesis Inhibitor 3 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB9125MI
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9125MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9125MI Supplier MilliporeSigma Supplier No. AB9125
Only null left

Rabbit Polyclonal Antibody

Anti-Brain-Specific Angiogenesis Inhibitor 3 Antibody is an antibody against Brain-Specific Angiogenesis Inhibitor 3 for use in immunohistochemistry (paraffin).

Specifications

Antigen Brain-Specific Angiogenesis Inhibitor 3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation Liquid in PBS, pH 7.7 with 0.01% sodium azide.
Gene Symbols BAI3; KIAA0550
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide from the 3rd cytoplasmic domain of human BAI3. The immunogen sequence has 35% similarity with mouse.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −70°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only