missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Blys/TALL-1/BAFF/THANK Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 07 167
Description
Anti-Blys/TALL-1/BAFF/THANK Antibody is an antibody against Blys/TALL-1/BAFF/THANK for use in western blotting.Specifications
| Blys/TALL-1/BAFF/THANK | |
| Polyclonal | |
| PBS containing 0.02% sodium azide | |
| Rabbit | |
| Protein G chromatography | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| TNFSF13B; DTL; TALL-1; ZTNF4; BLyS; THANK; TNFSF20; CD257; TALL1; BAFF; BLYS | |
| peptide corresponding to amino acids 254-269 of human Blys (EEGDELQLAIPRENAQ) | |
| 100 μg | |
| NM_006573 | |
| −20°C for 24 months | |
| IgG |
For Research Use Only