missing translation for 'onlineSavingsMsg'
Learn More

Bin1 Mouse anti-Human, Mouse, Rat, Clone: 99D, MilliporeSigma™

Catalog No. 05449MI
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05449MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05449MI Supplier MilliporeSigma Supplier No. 05449
Only null left

Mouse Monoclonal Antibody

Anti-Bin1 Antibody, clone 99D is an antibody against Bin1 for use in immunoprecipitation, western blot, immunohistochemistry.

Specifications

Antigen Bin1
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 99D
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, containing 0.05% sodium azide
Gene Accession No. NM_139343
Gene Symbols BIN1, AMPH2, amphiphysin-like, MGC10367, DKFZp547F068, AMPHL, SH3P9
Host Species Mouse
Immunogen Partial fusion protein corresponding to amino acids 189-398 of human Bin1. Clone 99D
Purification Method Protein G chromatography
Quantity 200 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only