missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Bikunin, Polyclonal, Invitrogen™
Supplier: METTLER TOLEDO 30667828
Description
Host Species: Rabbit
Species Reactivity: Human
Formulation: 200μL of Unpurified antibody in whole serum;
Immunogen: Human recombinant bikunin.
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=
ELISA and Western Blot
Specifications
| Bikunin | |
| Unconjugated | |
| Human recombinant bikunin. | |
| 200 μL | |
| Human |
| Polyclonal | |
| Rabbit | |
| Unpurified | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |