missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Atg18 (WIPI-2) Mouse anti-Human, Clone: 2A2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABC91
Description
This Anti-Atg18 (WIPI-2) Antibody, clone 2A2 is validated for use in western blot, immunohistochemistry (paraffin) for the detection of Atg18 (WIPI-2).Specifications
| Atg18 (WIPI-2) | |
| Monoclonal | |
| Unconjugated | |
| NP_001028690 | |
| Mouse | |
| Protein G Purified | |
| Primary | |
| 2-8°C for one year | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2A2 | |
| Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
| WIPI2 | |
| Linear peptide corresponding to the C-terminus of human Atg18. | |
| 100 μL | |
| Human | |
| Purified |
For Research Use Only