missing translation for 'onlineSavingsMsg'
Learn More

ASNA1, Mouse, Clone: 5A3, Abnova™

Mouse monoclonal antibody raised against a partial recombinant ASNA1.

Supplier:  Abnova Corporation H00000439M02

Catalog No. 89-016-227


Only null left
Explore available promotions

Description

Description

ASNA1 is the human homolog of the bacterial arsA gene. In E. coli, ArsA ATPase is the catalytic component of a multisubunit oxyanion pump that is responsible for resistance to arsenicals and antimonials.[supplied by OMIM

Sequence: PEQTTFICVCIAEFLSLYETERLIQELAKCKIDTHNIIVNQLVFPDPEKPCKMCEARHKIQAKYLDQMEDLYEDFHIVKLPLLPHEVRGADKVNTFSALLLEPYKPPSAQ
Specifications

Specifications

ASNA1
Monoclonal
Unconjugated
PBS with no preservative; pH 7.4
NM_004317
ASNA1
ASNA1 (NP_004308, 239 a.a. ∼ 348 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG1 κ
ELISA, Western Blot
5A3
Mouse monoclonal antibody raised against a partial recombinant ASNA1.
ASNA1
ARSA-I/ARSA1/MGC3821
Mouse
Affinity chromatography
RUO
439
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions