missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aquaporin 9 Chicken anti-Rat, Polyclonal, MilliporeSigma™
Chicken Polyclonal Antibody
Supplier: MilliporeSigma AB3091
Description
Detect Aquaporin 9 using this Anti-Aquaporin 9 Antibody validated for use in ELISA, immunocytochemistry, immunohistochemistry and western blotting.Specifications
| Aquaporin 9 | |
| Polyclonal | |
| Affinity Purified immunoglobulin. Liquid in PBS with 0.1% BSA. | |
| Chicken | |
| Affinity Purified | |
| Primary | |
| Rat |
| ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| AQP9; HsT17287; Aquaporin-9; AQP-9; SSC1 | |
| An 18 AA synthetic peptide within the C-terminal domain of rat Aquaporin-9 (Ishibashi et al. 1997) was selected for antibody production. This domain is predicted to be cytoplasmic. | |
| 50 μg | |
| NM_020980.3 | |
| −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles. |
For Research Use Only