missing translation for 'onlineSavingsMsg'
Learn More

APP-C99 Mouse anti-Human, Mouse, Clone: mC99(1-7), MilliporeSigma™

Catalog No. MABN381MI
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN381MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN381MI Supplier MilliporeSigma Supplier No. MABN381
Only null left

Mouse Monoclonal Antibody

Anti-APP-C99 Antibody, clone mC99(1-7) is a highly specific mouse monoclonal antibody, that targets APP and has been tested in western blotting, immunoprecipitation, immunocytochemistry and immunohistochemistry.

Specifications

Antigen APP-C99
Applications Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone mC99(1-7)
Conjugate Unconjugated
Formulation Mouse monoclonal IgGκ in ascites containing 0.05% sodium azide.
Gene Accession No. NP_000475
Gene Symbols APP, A4, AD1
Host Species Mouse
Immunogen Recombinant protein corresponding to Human APP-C99.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Show More Show Less

For Research Use Only