missing translation for 'onlineSavingsMsg'
Learn More

APC Mouse anti-Human, Mouse, Clone: C-APC 28.9, MilliporeSigma™

Catalog No. MAB3786MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3786MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3786MI Supplier MilliporeSigma Supplier No. MAB3786

Mouse Monoclonal Antibody

Anti-APC Antibody, C-terminus, clone C-APC 28.9 is a Mouse Monoclonal Antibody for detection of APC also known as Adenomatous Polyposis Coli Protein and has been validated in immunofluoresence, immunoprecipitation, western blotting and immunohistochemistry.

Specifications

Antigen APC
Applications Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone C-APC 28.9
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.02M phosphate buffer, 0.25M NaCl, pH 7.6, with 0.1% sodium azide.
Gene Accession No. NM_000038.3
Gene Symbols APC, DP2.5, FAP, GS, DP3, DP2, FPC
Host Species Mouse
Immunogen Human APC C-terminal fusion with MBP (maltose binding protein).
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only