missing translation for 'onlineSavingsMsg'
Learn More

Ambra1 Rabbit anti-Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. abc131mi
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABC131MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABC131MI Supplier MilliporeSigma Supplier No. ABC131
Only null left

Rabbit Polyclonal Antibody

Anti-Ambra1 Antibody is a rabbit polyclonal antibody, validated for use in western blotting.

Specifications

Antigen Ambra1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols AMBRA1
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to the C-terminus of mouse Ambra1.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_7662587
Target Species Human, Mouse
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only