missing translation for 'onlineSavingsMsg'
Learn More

Alzheimer Precursor Protein A4 (NT) Mouse anti-Canine, FITC, Clone: 22C11, MilliporeSigma™

Catalog No. fcmab327fmi
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB327FMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB327FMI Supplier MilliporeSigma Supplier No. FCMAB327F

Mouse Monoclonal Antibody

Anti-Alzheimer Precursor Protein A4 (N-terminus)-FITC Antibody, clone 22C11 detects level of Alzheimer Precursor Protein A4 (N-terminus) and has been published and validated for use in flow cytometry.

Specifications

Antigen Alzheimer Precursor Protein A4 (NT)
Applications Flow Cytometry
Classification Monoclonal
Clone 22C11
Conjugate FITC
Formulation Purified mouse monoclonal IgG1 conjugated to FITC in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Accession No. NP_000475
Gene Symbols APP, PN-II, AD1, ABETA, CTFgamma, APPI, ABPP, A4, CVAP, PN2, PreA4, AAA
Host Species Mouse
Immunogen Purified recombinant Alzheimer precursor A4 (pre A4695) fusion protein.
Purification Method Protein A Purified
Quantity 100 Tests
Primary or Secondary Primary
Target Species Canine
Content And Storage 2-8°C for up to 6 months. Protect from light.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only