Learn More
ALAS1, Mouse, Clone: 2D5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ALAS1.
Supplier: Abnova Corporation H00000211M03
Description
Delta-aminolevulinate synthase (ALAS; EC 2.3.1.37) catalyzes the condensation of glycine with succinyl-CoA to form delta-aminolevulinic acid. This nuclear-encoded mitochondrial enzyme is the first and rate-limiting enzyme in the mammalian heme biosynthetic pathway. There are 2 tissue-specific isozymes: a housekeeping enzyme encoded by the ALAS1 gene and an erythroid tissue-specific enzyme encoded by ALAS2 (MIM 301300).[supplied by OMIM
Sequence: MESVVRRCPFLSRVPQAFLQKAGKSLLFYAQNCPKMMEVGAKPAPRALSTAAVHYQQIKETPPASEKDKTAKAKVQQTPDGSQQSPDGTQLPSGHPLPSpecifications
| ALAS1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_000688 | |
| ALAS1 | |
| ALAS1 (NP_000679, 1 a.a. ∼ 98 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2D5 | |
| Mouse monoclonal antibody raised against a partial recombinant ALAS1. | |
| ALAS1 | |
| ALAS/ALAS3/ALASH/MIG4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 211 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |