Learn More
v-akt murine thymoma viral oncogene homolog 2, Mouse, Clone: 1B4, Abnova™
Mouse monoclonal antibody raised against a partial recombinant AKT2.
Supplier: Abnova Corporation H00000208M02
Description
This gene is a putative oncogene encoding a protein belonging to a subfamily of serine/threonine kinases containing SH2-like (Src homology 2-like) domains. The gene was shown to be amplified and overexpressed in 2 of 8 ovarian carcinoma cell lines and 2 of 15 primary ovarian tumors. Overexpression contributes to the malignant phenotype of a subset of human ductal pancreatic cancers. The encoded protein is a general protein kinase capable of phophorylating several known proteins. [provided by RefSeq
Sequence: MRAIQMVANSLKQRAPGEDPMDYKCGSPSDSSTTEEMEVAVSKARAKVTMNDFDYLKLLGKGTFGKVILVREKATGRYYAMKILRKEVIISpecifications
| v-akt murine thymoma viral oncogene homolog 2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| M95936 | |
| AKT2 | |
| AKT2 (AAA58364, 100 a.a. ∼ 189 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Kinases | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody |
| ELISA, Western Blot | |
| 1B4 | |
| Mouse monoclonal antibody raised against a partial recombinant AKT2. | |
| AKT2 | |
| PKBB/PKBBETA/PRKBB/RAC-BETA | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 208 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |