missing translation for 'onlineSavingsMsg'
Learn More

AKAP 150 Rabbit anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. 07210MI
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07210MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07210MI Supplier MilliporeSigma Supplier No. 07210
Only null left

Rabbit Polyclonal Antibody

Anti-AKAP 150 Antibody is an antibody against AKAP 150 for use in western blotting.

Specifications

Antigen AKAP 150
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Gene Symbols AKAP1; AKAP121; AKAP84-PEN; SAKAP84; PRKA1; D-AKAP-1; AKAP; D-AKAP1; AKAP149; MGC1807; AKAP84; S-AKAP84
Host Species Rabbit
Immunogen peptide corresponding to amino acids 428-449 of rat AKAP 150 (C-TTVGQAEEATVGQAEEATVGQA)
Purification Method Antiserum
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_598199
Target Species Rat
Content And Storage −20°C for 24 months
Form Serum
Isotype IgG
Show More Show Less

For Research Use Only