Learn More
ADAM2 Mouse anti-Mouse, Clone: 9D2.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier:  MilliporeSigma MAB19292
Catalog No. MAB19292MI
Only null left
Explore available promotions
Description
Description
This Anti-ADAM2 Antibody, clone 9D2.2 is validated for use in ELISA, western blotting for the detection of ADAM2.
Specifications
Specifications
ADAM2
Monoclonal
Unconjugated
NM_001464.3
Mouse
100 μg
Mouse
Purified
ELISA, Western Blot
9D2.2
Liquid at 1mg/mL in PBS containing 0.05% sodium azide.
ADAM2, PH30, CRYN1, FTNB, PH30-beta, CRYN2, PH-30b, PH-30, CT15
Recombinant mouse fertilin produced by baculovirus expressin in SF9 cells.
Primary
2-8°C in undiluted aliquots for up to 12 months.
IgG2b
Product Suggestions
Product Suggestions