missing translation for 'onlineSavingsMsg'
Learn More

anti-Actin, Clone: C4, MP Biomedicals

Catalog No. ICN691001
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN691001 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN691001 Supplier MP Biomedicals Inc Supplier No. 08691001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Host Species: Mouse
Clone: C4
Isotype: IgG
Species Reactivity: Human

Specifications

Antigen Actin
Classification Monoclonal
Clone C4
Conjugate Unconjugated
Host Species Mouse
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less